siRNA Details
| Gene Symbol: Nudt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181363.2 | NP_852028.1 | 5 | 1016 | ||
| XM_008760905.1 | XP_008759127.1 | 5 | 822 | |||
| GenBank mRNA | AF188995.1 | - | 4 | 791 | ||
| AF188996.1 | - | 3 | 735 | |||
| U58289.1 | - | 5 | 1016 | |||

Nudt6
Genomic Map