siRNA Details
| Gene Symbol: Kcnip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181365.2 | NP_852030.1 | 1195 | |||
| GenBank mRNA | AF345444.1 | - | 1195 | |||
| AF345445.1 | - | 7 | 765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnip4| Gene Symbol: Kcnip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181365.2 | NP_852030.1 | 1195 | |||
| GenBank mRNA | AF345444.1 | - | 1195 | |||
| AF345445.1 | - | 7 | 765 | |||