siRNA Details
| Gene Symbol: Lhx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181367.2 | NP_852032.1 | 4, 5 | 470 | ||
| XM_006249914.2 | XP_006249976.1 | 2, 3 | 962 | |||
| XM_006249916.2 | XP_006249978.1 | 5, 6 | 1032 | |||
| XM_006249917.1 | XP_006249979.1 | 2, 3 | 323 | |||
| XM_006249918.2 | XP_006249980.1 | 2, 3 | 957 | |||
| XM_006249919.2 | XP_006249981.1 | 5, 6 | 1032 | |||
| XM_008769545.1 | XP_008767767.1 | 4, 5 | 558 | |||
| XM_008769546.1 | XP_008767768.1 | 2, 3 | 957 | |||
| GenBank mRNA | AF527619.1 | - | 2, 3 | 278 | ||
| AY273890.1 | - | 4, 5 | 470 | |||
| AY724502.1 | - | 4, 5 | 470 | |||
| BC128722.1 | - | 5, 6 | 546 | 8362986 | ||

Lhx9
Genomic Map