siRNA Details
| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181382.2 | NP_852047.1 | 10 | 1402 | ||
| GenBank mRNA | AY266116.1 | - | 10 | 1402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gldn| Gene Symbol: Gldn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181382.2 | NP_852047.1 | 10 | 1402 | ||
| GenBank mRNA | AY266116.1 | - | 10 | 1402 | ||