siRNA Details
| Gene Symbol: WDR45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029896.1 | NP_001025067.1 | 8 | 819 | ||
| NM_007075.3 | NP_009006.2 | 9 | 1089 | |||
| GenBank mRNA | AJ005897.1 | - | 8 | 775 | ||
| AK023344.1 | - | 6 | 1797 | |||
| AK225122.1 | - | 8 | 798 | |||
| AK315717.1 | - | 8 | 806 | |||
| AY691428.1 | - | 7 | 647 | |||
| BC000464.2 | - | 8 | 813 | 2822742 | ||
| BC003037.1 | - | 8 | 813 | 2822742 | ||
| BC009027.1 | - | 5 | 476 | |||
| BC069206.1 | - | 9 | 1089 | 5801768 | ||

WDR45
Genomic Map