siRNA Details
| Gene Symbol: Niacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181476.1 | NP_852141.1 | 1 | 184 | ||
| GenBank mRNA | AB103062.1 | - | 1 | 184 | ||
More Information
Other Species Targeted
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Niacr1| Gene Symbol: Niacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181476.1 | NP_852141.1 | 1 | 184 | ||
| GenBank mRNA | AB103062.1 | - | 1 | 184 | ||
Set Membership: |
|