siRNA Details
Gene Symbol: Rdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181478.2 | NP_852143.1 | 2 | 355 | ||
GenBank mRNA | AY178865.1 | - | 2 | 355 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181478.2 | NP_852143.1 | 2 | 355 | ||
GenBank mRNA | AY178865.1 | - | 2 | 355 |