siRNA Details
| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181478.2 | NP_852143.1 | 6 | 1013 | ||
| GenBank mRNA | AY178865.1 | - | 6 | 1013 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh10| Gene Symbol: Rdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181478.2 | NP_852143.1 | 6 | 1013 | ||
| GenBank mRNA | AY178865.1 | - | 6 | 1013 | ||