siRNA Details
| Gene Symbol: Kif15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181635.2 | NP_853666.1 | 3 | 240 | ||
| GenBank mRNA | AY291580.1 | - | 3 | 240 | ||
| AY291581.1 | - | 3 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif15| Gene Symbol: Kif15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181635.2 | NP_853666.1 | 3 | 240 | ||
| GenBank mRNA | AY291580.1 | - | 3 | 240 | ||
| AY291581.1 | - | 3 | 240 | |||