siRNA Details
| Gene Symbol: Slc29a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181639.3 | NP_853670.2 | 2 | 230 | ||
| GenBank mRNA | AY273196.1 | - | 1 | 152 | ||
| BC078678.1 | - | 2 | 230 | 7132846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc29a3| Gene Symbol: Slc29a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181639.3 | NP_853670.2 | 2 | 230 | ||
| GenBank mRNA | AY273196.1 | - | 1 | 152 | ||
| BC078678.1 | - | 2 | 230 | 7132846 | ||