siRNA Details
| Gene Symbol: Kiss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181692.1 | NP_859043.1 | 2 | 333 | ||
| XM_008769443.1 | XP_008767665.1 | 3 | 786 | |||
| GenBank mRNA | AY196983.1 | - | 2 | 333 | ||
| JX139030.1 | - | 3 | 744 | |||
| JX139031.1 | - | 3 | 540 | |||
| JX139032.1 | - | 3 | 465 | |||

Kiss1
Genomic Map