siRNA Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082580.1 | NP_001076049.1 | 3 | 395 | ||
| NM_182668.2 | NP_872609.2 | 3 | 363 | |||
| GenBank mRNA | AY300752.1 | - | 2 | 226 | ||
| BC087146.1 | - | 3 | 344 | 7129801 | ||
| CB576182.1 | - | 3 | 458 | |||
| CO558072.1 | - | 3 | 401 | |||
| FQ213884.1 | - | 250 | ||||
| FQ227281.1 | - | 392 | ||||

Nit1
Genomic Map