siRNA Details
| Gene Symbol: Micalcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182669.2 | NP_872610.2 | 4 | 1364 | ||
| GenBank mRNA | AY149342.1 | - | 1 | 804 | ||
| BC098647.1 | - | 4 | 1364 | 7112444 | ||
| BC100137.1 | - | 1 | 815 | 7457561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Micalcl| Gene Symbol: Micalcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182669.2 | NP_872610.2 | 4 | 1364 | ||
| GenBank mRNA | AY149342.1 | - | 1 | 804 | ||
| BC098647.1 | - | 4 | 1364 | 7112444 | ||
| BC100137.1 | - | 1 | 815 | 7457561 | ||