siRNA Details
Gene Symbol: Ccdc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182736.1 | NP_877400.1 | 3 | 125 | ||
XM_008768795.1 | XP_008767017.1 | 3 | 368 | |||
GenBank mRNA | AJ534984.1 | - | 3 | 125 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182736.1 | NP_877400.1 | 3 | 125 | ||
XM_008768795.1 | XP_008767017.1 | 3 | 368 | |||
GenBank mRNA | AJ534984.1 | - | 3 | 125 |