siRNA Details
| Gene Symbol: Tmem255a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182822.3 | NP_877974.2 | 2, 3 | 392 | ||
| XM_006257479.2 | XP_006257541.1 | 2, 3 | 392 | |||
| GenBank mRNA | AY327410.1 | - | 2, 3 | 257 | ||
| CB580315.1 | - | 2, 3 | 320 | |||

Tmem255a
Genomic Map