siRNA Details
| Gene Symbol: Cd276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182824.2 | NP_877976.1 | 8 | 2041 | ||
| GenBank mRNA | AY190319.1 | - | 8 | 2043 | ||
| BC100064.1 | - | 8 | 2041 | 7104832 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd276| Gene Symbol: Cd276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182824.2 | NP_877976.1 | 8 | 2041 | ||
| GenBank mRNA | AY190319.1 | - | 8 | 2043 | ||
| BC100064.1 | - | 8 | 2041 | 7104832 | ||