siRNA Details
| Gene Symbol: Rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182825.1 | NP_877977.1 | 3, 4 | 382 | ||
| XM_008765754.1 | XP_008763976.1 | 3, 4 | 400 | |||
| XM_008765755.1 | XP_008763977.1 | 3, 4 | 400 | |||
| GenBank mRNA | AY327411.1 | - | 3, 4 | 382 | ||

Rhebl1
Genomic Map