siRNA Details
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182825.1 | NP_877977.1 | 8 | 912 | ||
XM_008765754.1 | XP_008763976.1 | 8 | 907 | |||
GenBank mRNA | AY327411.1 | - | 8 | 912 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182825.1 | NP_877977.1 | 8 | 912 | ||
XM_008765754.1 | XP_008763976.1 | 8 | 907 | |||
GenBank mRNA | AY327411.1 | - | 8 | 912 |