siRNA Details
| Gene Symbol: Camk1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182842.1 | NP_878262.1 | 3 | 211 | ||
| XM_006250470.2 | XP_006250532.1 | 3 | 328 | |||
| GenBank mRNA | AB101231.1 | - | 3 | 211 | ||
| AB101232.1 | - | 3 | 211 | |||
| D86557.1 | - | 3 | 217 | |||

Camk1g
Genomic Map