siRNA Details
| Gene Symbol: Ston1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182954.2 | NP_891999.2 | 2 | 875 | ||
| XM_006239743.2 | XP_006239805.1 | 2 | 646 | |||
| XM_006239744.2 | XP_006239806.1 | 2 | 1742 | |||
| XM_008764479.1 | XP_008762701.1 | 3 | 963 | |||
| XM_008764480.1 | XP_008762702.1 | 2 | 1431 | |||
| XM_008764481.1 | XP_008762703.1 | 2 | 626 | |||
| GenBank mRNA | AY300793.1 | - | 1 | 593 | ||

Ston1
Genomic Map