siRNA Details
| Gene Symbol: Ube2v2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183052.2 | NP_898875.1 | 2 | 113 | ||
| XM_006248727.2 | XP_006248789.1 | 2 | 147 | |||
| GenBank mRNA | AJ515244.1 | - | 2 | 101 | ||
| BC087593.1 | - | 2 | 91 | 7310168 | ||
| FM082155.1 | - | 2 | 103 | |||
| FQ216277.1 | - | 2 | 94 | |||

Ube2v2
Genomic Map