siRNA Details
| Gene Symbol: Potea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770390.1 | XP_008768612.1 | 5 | 953 | ||
| XM_008770391.1 | XP_008768613.1 | 5 | 953 | |||
| XM_008770392.1 | XP_008768614.1 | 4 | 779 | |||
| GenBank mRNA | AB095021.1 | - | 5 | 822 | ||
| BC083629.1 | - | 5 | 927 | 7134382 | ||

Potea
Genomic Map