siRNA Details
| Gene Symbol: Psmd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198730.2 | NP_942025.1 | 2 | 373 | ||
| GenBank mRNA | BC059159.1 | - | 2 | 378 | 6887177 | |
| FQ212955.1 | - | 2 | 373 | |||
| FQ213157.1 | - | 2 | 405 | |||
| FQ213627.1 | - | 2 | 397 | |||
| FQ214459.1 | - | 2 | 373 | |||
| FQ215751.1 | - | 2 | 373 | |||
| FQ229068.1 | - | 2 | 390 | |||

Psmd6
Genomic Map