siRNA Details
| Gene Symbol: Chdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198731.2 | NP_942026.1 | 8 | 1675 | ||
| XM_006252578.2 | XP_006252640.1 | 8 | 1740 | |||
| XM_006252579.2 | XP_006252641.1 | 8 | 1694 | |||
| XM_006252580.2 | XP_006252642.1 | 8 | 1500 | |||
| XM_008770984.1 | XP_008769206.1 | 8 | 2043 | |||
| XM_008770985.1 | XP_008769207.1 | 9 | 2132 | |||
| GenBank mRNA | AY365023.1 | - | 7 | 1424 | ||
| BC085787.1 | - | 8 | 1675 | 7108667 | ||
| X94769.1 | - | 7 | 950 | |||

Chdh
Genomic Map