siRNA Details
| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198736.1 | NP_942031.1 | 6 | 973 | ||
| GenBank mRNA | AY305029.1 | - | 6 | 973 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd13| Gene Symbol: Kctd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198736.1 | NP_942031.1 | 6 | 973 | ||
| GenBank mRNA | AY305029.1 | - | 6 | 973 | ||