siRNA Details
Gene Symbol: Rgsl2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198790.1 | NP_942085.1 | 1 | 190 | ||
GenBank mRNA | AJ515383.1 | - | 1 | 190 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rgsl2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198790.1 | NP_942085.1 | 1 | 190 | ||
GenBank mRNA | AJ515383.1 | - | 1 | 190 |