siRNA Details
| Gene Symbol: Nme8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198792.2 | NP_942087.2 | 6 | 434 | ||
| XM_006254088.2 | XP_006254150.1 | 6 | 483 | |||
| GenBank mRNA | AF548544.1 | - | 5 | 330 | ||
| BC097265.1 | - | 6 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nme8| Gene Symbol: Nme8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198792.2 | NP_942087.2 | 6 | 434 | ||
| XM_006254088.2 | XP_006254150.1 | 6 | 483 | |||
| GenBank mRNA | AF548544.1 | - | 5 | 330 | ||
| BC097265.1 | - | 6 | 428 | |||