siRNA Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012504.1 | NP_036636.1 | 4, 5 | 618 | ||
| GenBank mRNA | BC061968.1 | - | 4, 5 | 629 | 5598014 | |
| M14511.1 | - | 4, 5 | 618 | |||
| M28647.1 | - | 4, 5 | 641 | |||
| X05882.1 | - | 4, 5 | 395 | |||

Atp1a1
Genomic Map