siRNA Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012505.2 | NP_036637.1 | 3 | 265 | ||
| GenBank mRNA | BC085764.1 | - | 3 | 265 | 7190910 | |
| M14512.1 | - | 3 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp1a2| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012505.2 | NP_036637.1 | 3 | 265 | ||
| GenBank mRNA | BC085764.1 | - | 3 | 265 | 7190910 | |
| M14512.1 | - | 3 | 250 | |||