siRNA Details
| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012640.2 | NP_036772.2 | 2 | 201 | ||
| XM_006243607.1 | XP_006243669.1 | 3 | 381 | |||
| GenBank mRNA | M13949.1 | - | 2 | 201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbp2| Gene Symbol: Rbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012640.2 | NP_036772.2 | 2 | 201 | ||
| XM_006243607.1 | XP_006243669.1 | 3 | 381 | |||
| GenBank mRNA | M13949.1 | - | 2 | 201 | ||