siRNA Details
| Gene Symbol: Acot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146061.1 | NP_001139533.1 | 2 | 282 | ||
| NM_013214.4 | NP_037346.2 | 2 | 210 | |||
| XM_006239447.2 | XP_006239509.1 | 2 | 310 | |||
| XM_006239448.1 | XP_006239510.1 | 2 | 218 | |||
| GenBank mRNA | BC087716.1 | - | 2 | 162 | 7316866 | |
| CK469690.1 | - | 235 | ||||
| D88891.1 | - | 2 | 282 | |||
| DY317682.1 | - | 2 | 294 | |||
| Y09332.1 | - | 2 | 110 | |||

Acot7
Genomic Map