siRNA Details
| Gene Symbol: Hbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013221.2 | NP_037353.2 | 9 | 1228 | ||
| XM_006239975.2 | XP_006240037.1 | 9 | 1291 | |||
| GenBank mRNA | BC091165.1 | - | 8 | 1202 | 7097013 | |
| FQ223257.1 | - | 9 | 1335 | |||
| U09551.1 | - | 8 | 1223 | |||

Hbp1
Genomic Map