siRNA Details
| Gene Symbol: Aldh1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017272.15 | NP_058968.14 | 8 | 950 | ||
| XM_008760315.1 | XP_008758537.1 | 8 | 929 | |||
| GenBank mRNA | M23995.1 | - | 8 | 950 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh1a7| Gene Symbol: Aldh1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017272.15 | NP_058968.14 | 8 | 950 | ||
| XM_008760315.1 | XP_008758537.1 | 8 | 929 | |||
| GenBank mRNA | M23995.1 | - | 8 | 950 | ||