siRNA Details
| Gene Symbol: Aco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017321.1 | NP_059017.1 | 3 | 454 | ||
| GenBank mRNA | L23874.1 | - | 3 | 454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aco1| Gene Symbol: Aco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017321.1 | NP_059017.1 | 3 | 454 | ||
| GenBank mRNA | L23874.1 | - | 3 | 454 | ||