siRNA Details
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017335.1 | NP_059031.1 | 3 | 605 | ||
GenBank mRNA | FQ229573.1 | - | 129 | |||
U28927.1 | - | 3 | 605 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017335.1 | NP_059031.1 | 3 | 605 | ||
GenBank mRNA | FQ229573.1 | - | 129 | |||
U28927.1 | - | 3 | 605 |