siRNA Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017340.2 | NP_059036.1 | 224 | |||
| GenBank mRNA | BC085743.1 | - | 224 | 7109722 | ||
| J02752.1 | - | 207 | ||||
| Gene Symbol: LOC100910385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750965.2 | XP_003751013.1 | 2 | 218 | ||

Acox1
Genomic Map