siRNA Details
| Gene Symbol: Itih3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017351.1 | NP_059047.1 | 9, 10 | 1088 | ||
| XM_006252649.1 | XP_006252711.1 | 9, 10 | 1318 | |||
| XM_006252650.1 | XP_006252712.1 | 9, 10 | 1318 | |||
| XM_006252653.1 | XP_006252715.1 | 9, 10 | 1318 | |||
| XM_008771002.1 | XP_008769224.1 | 9, 10 | 1318 | |||
| GenBank mRNA | X83231.1 | - | 9, 10 | 1088 | ||

Itih3
Genomic Map