siRNA Details
| Gene Symbol: Enpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019370.2 | NP_062243.2 | 2, 3 | 206 | ||
| GenBank mRNA | BC097326.1 | - | 2, 3 | 206 | 7458431 | |
| D30649.1 | - | 2, 3 | 233 | |||
| U78787.1 | - | 2, 3 | 203 | |||
| U78788.1 | - | 2, 3 | 209 | |||
| Z47987.1 | - | 2, 3 | 190 | |||

Enpp3
Genomic Map