siRNA Details
| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019379.1 | NP_062252.1 | 3 | 285 | ||
| XM_006250750.2 | XP_006250812.1 | 3 | 306 | |||
| GenBank mRNA | U14192.1 | - | 3 | 200 | ||
| U15589.1 | - | 3 | 285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uso1| Gene Symbol: Uso1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019379.1 | NP_062252.1 | 3 | 285 | ||
| XM_006250750.2 | XP_006250812.1 | 3 | 306 | |||
| GenBank mRNA | U14192.1 | - | 3 | 200 | ||
| U15589.1 | - | 3 | 285 | |||