siRNA Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019907.1 | NP_063972.1 | 4 | 272 | ||
| GenBank mRNA | AF047384.1 | - | 4 | 272 | ||
| FQ212096.1 | - | 1 | 17 | |||
| FQ222669.1 | - | 4 | 305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cript| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019907.1 | NP_063972.1 | 4 | 272 | ||
| GenBank mRNA | AF047384.1 | - | 4 | 272 | ||
| FQ212096.1 | - | 1 | 17 | |||
| FQ222669.1 | - | 4 | 305 | |||