siRNA Details
| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020091.1 | NP_064476.1 | 2 | 210 | ||
| GenBank mRNA | AJ251688.1 | - | 2 | 210 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cym| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020091.1 | NP_064476.1 | 2 | 210 | ||
| GenBank mRNA | AJ251688.1 | - | 2 | 210 | ||