siRNA Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020092.1 | NP_064477.1 | 2 | 203 | ||
| XM_006231892.2 | XP_006231954.1 | 4 | 722 | |||
| XM_006231894.2 | XP_006231956.1 | 3 | 5131 | |||
| GenBank mRNA | AF234765.1 | - | 2 | 203 | ||

Srek1
Genomic Map