siRNA Details
| Gene Symbol: Ccr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020542.2 | NP_065417.1 | 2 | 564 | ||
| GenBank mRNA | BC079207.1 | - | 2 | 564 | 7131790 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr1| Gene Symbol: Ccr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020542.2 | NP_065417.1 | 2 | 564 | ||
| GenBank mRNA | BC079207.1 | - | 2 | 564 | 7131790 | |