siRNA Details
| Gene Symbol: Tmem33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034198.2 | NP_001029370.1 | 4 | 323 | ||
| NM_021671.3 | NP_067703.1 | 4 | 323 | |||
| GenBank mRNA | AB006135.1 | - | 4 | 281 | ||
| CK359233.1 | - | 4 | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem33| Gene Symbol: Tmem33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034198.2 | NP_001029370.1 | 4 | 323 | ||
| NM_021671.3 | NP_067703.1 | 4 | 323 | |||
| GenBank mRNA | AB006135.1 | - | 4 | 281 | ||
| CK359233.1 | - | 4 | 204 | |||