siRNA Details
Gene Symbol: Tmem33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034198.2 | NP_001029370.1 | 4 | 326 | ||
NM_021671.3 | NP_067703.1 | 4 | 326 | |||
GenBank mRNA | AB006135.1 | - | 4 | 284 | ||
CK359233.1 | - | 4 | 207 |
