siRNA Details
| Gene Symbol: Negr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021682.1 | NP_067714.1 | 2 | 467 | ||
| GenBank mRNA | AB017139.1 | - | 2 | 467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Negr1| Gene Symbol: Negr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021682.1 | NP_067714.1 | 2 | 467 | ||
| GenBank mRNA | AB017139.1 | - | 2 | 467 | ||