siRNA Details
| Gene Symbol: Cnksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113366.1 | NP_001106837.1 | 17, 18 | 2157 | ||
| NM_021686.3 | NP_067718.1 | 17, 18 | 2157 | |||
| XM_008773224.1 | XP_008771446.1 | 16, 17 | 2226 | |||
| GenBank mRNA | AF102853.1 | - | 17, 18 | 1984 | ||
| AF102854.1 | - | 17, 18 | 2446 | |||

Cnksr2
Genomic Map