siRNA Details
| Gene Symbol: Erbb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021687.1 | NP_067719.1 | 3, 4 | 455 | ||
| GenBank mRNA | AF041838.1 | - | 3, 4 | 455 | ||
| AY375306.1 | - | 3, 4 | 422 | |||
| AY375307.1 | - | 3, 4 | 422 | |||
| AY375308.1 | - | 3, 4 | 422 | |||

Erbb4
Genomic Map