siRNA Details
| Gene Symbol: Rapgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021690.1 | NP_067722.1 | 3 | 356 | ||
| XM_006242352.2 | XP_006242414.1 | 4 | 616 | |||
| XM_008765778.1 | XP_008764000.1 | 4 | 905 | |||
| XM_008765779.1 | XP_008764001.1 | 4 | 523 | |||
| XM_008765780.1 | XP_008764002.1 | 3 | 863 | |||
| GenBank mRNA | BC107667.1 | - | 3 | 309 | 7936411 | |
| U78167.1 | - | 3 | 356 | |||

Rapgef3
Genomic Map