siRNA Details
| Gene Symbol: Rbck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021764.1 | NP_068532.2 | 9 | 1391 | ||
| XM_006235252.2 | XP_006235314.1 | 10 | 1316 | |||
| GenBank mRNA | AB011369.1 | - | 8 | 1230 | ||
| BC061723.1 | - | 9 | 1325 | 5598480 | ||
| U48248.1 | - | 9 | 1391 | |||

Rbck1
Genomic Map